Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricyclohexylphosphine, 98%

$135.16 - $135.16

Chemical Identifiers

Chemical Name or Material Tricyclohexylphosphine
CAS 2622-14-2
Formula Weight 280.42
MDL Number MFCD00003853
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4195206

  SDS
alfa aesar™
4195206
5g Each for $135.16
Description & Specifications

Specifications

Formula Weight 280.42
Chemical Name or Material Tricyclohexylphosphine
TSCA TSCA
Physical Form Crystalline
Assay 98%
MDL Number MFCD00003853
CAS 2622-14-2
Health Hazard 1 Warning
Odor Unpleasant
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 76° to 78°C
Color White to off-white
Sensitivity Air Sensitive
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.