missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tricyclo[6.4.0.0(2,7)]dodecane-1,8:2,7-tetracarboxylic Dianhydride 98.0+%, TCI America™
$474.48 - $1644.06
Chemical Identifiers
CAS | 738-90-9 |
---|---|
Molecular Formula | C16H16O6 |
Molecular Weight (g/mol) | 304.30 |
MDL Number | MFCD11055168 |
InChI Key | YRPOEHNJJZOQJH-UHFFFAOYSA-N |
Synonym | Octahydrobiphenylene-4a,8b:4b,8a-tetracarboxylic Dianhydride, Octahydro-4a,8b:4b,8a-bis(methanooxymethano)biphenylene-9,11,12,14-tetraone |
PubChem CID | 53384377 |
IUPAC Name | 14,17-dioxapentacyclo[6.4.3.3²,â·.0¹,â¸.0²,â·]octadecane-13,15,16,18-tetrone |
SMILES | O=C1OC(=O)C23CCCCC12C12CCCCC31C(=O)OC2=O |
Chemical Identifiers
738-90-9 | |
304.30 | |
YRPOEHNJJZOQJH-UHFFFAOYSA-N | |
53384377 | |
O=C1OC(=O)C23CCCCC12C12CCCCC31C(=O)OC2=O |
C16H16O6 | |
MFCD11055168 | |
Octahydrobiphenylene-4a,8b:4b,8a-tetracarboxylic Dianhydride, Octahydro-4a,8b:4b,8a-bis(methanooxymethano)biphenylene-9,11,12,14-tetraone | |
14,17-dioxapentacyclo[6.4.3.3²,â·.0¹,â¸.0²,â·]octadecane-13,15,16,18-tetrone |
Specifications
738-90-9 | |
White-Yellow | |
MFCD11055168 | |
Octahydrobiphenylene-4a,8b:4b,8a-tetracarboxylic Dianhydride, Octahydro-4a,8b:4b,8a-bis(methanooxymethano)biphenylene-9,11,12,14-tetraone | |
O=C1OC(=O)C23CCCCC12C12CCCCC31C(=O)OC2=O | |
304.30 | |
304.30 | |
Crystalline Powder |
283°C | |
C16H16O6 | |
5 g | |
YRPOEHNJJZOQJH-UHFFFAOYSA-N | |
14,17-dioxapentacyclo[6.4.3.3²,â·.0¹,â¸.0²,â·]octadecane-13,15,16,18-tetrone | |
53384377 | |
≥98.0% (GC) | |
Tricyclo[6.4.0.0(2,7)]dodecane-1,8:2,7-tetracarboxylic Dianhydride |
Safety and Handling
TSCA : No