Learn More

Tricine (White Crystals), Fisher BioReagents
Manufacturer: Fisher BioReagents BP315100
Description
This biological buffer has a usable pH range of 7.4 to 8.8.Specifications
At 260nm (0.1M solution) is ≤0.04 At 280nm (0.1M solution) is ≤0.02 | |
Pass Test | |
White crystals | |
C6H13NO5 | |
15, 9816 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
79784 | |
179.09 | |
Powder/Solid | |
Pass test | |
Poly Bottle | |
182°C | |
Undesignated |
Tricine | |
5704-04-1 | |
≥98 % | |
(HOCH2)3CNHCH2CO2H | |
N-[Tris-(hydroxymethyl)methyl]glycine | |
C(C(=O)O)NC(CO)(CO)CO | |
179.17 | |
CHEBI:39063 | |
Pass Test | |
≥98% | |
≤1.0% | |
7.9 to 8.3 (at 25°C) | |
4.6 to 6.0 | |
100g |
Chemical Identifiers
5704-04-1 | |
179.17 | |
N-[Tris-(hydroxymethyl)methyl]glycine | |
CHEBI:39063 | |
C(C(=O)O)NC(CO)(CO)CO |
C6H13NO5 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
79784 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
Safety and Handling

WARNING!
Emergency Overview
Causes eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:2
Flammability:1
Instability:0
Recommended Storage : RT
Promotions
-
Fisher BioReagents
Get 2 Extra Life Sciences Chemicals When You Buy 3
Buy three life sciences chemicals and get two more at no additional cost.: 03/31/21