missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricine, 0.5M buffer soln., pH 7.0

$59.51 - $93.45

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62234AE View Documents Alfa Aesar™
J62234AE
100mL Each for $59.51
 
AAJ62234AK View Documents Alfa Aesar™
J62234AK
250mL Each for $93.45
 
Specifications

Chemical Identifiers

5704-04-1
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO
C6H13NO5
MFCD00004277
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid

Specifications

5704-04-1
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
Liquid
Colorless
Tricine buffer soln.
C6H13NO5
0.5M buffer soln.
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
7.0
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA