Please login to your online account to display your discounted pricing

Trichlorovinylsilane, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trichlorovinylsilane
Name Note 96%
CAS 75-94-5
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C2H3Cl3Si
Linear Formula H2C=CHSiCl3
Formula Weight 161.49
MDL Number MFCD00000482
Synonym VTC
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147420100

  SDS
thermo fisher scientific
147420100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 161.49
Quantity 10mL
Color Clear colorless to slightly brown
EINECS Number 200-917-8
Density 1.2700g/mL
TSCA TSCA
RTECS Number VV6125000
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Synonym VTC
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS 75-94-5
Linear Formula H2C=CHSiCl3
Molecular Formula C2H3Cl3Si
Assay Percent Range 96%
Name Note 96%
Chemical Name or Material Trichlorovinylsilane
MDL Number MFCD00000482
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.