Please login to your online account to display your discounted pricing

Trichlorovinylsilane, 96%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC147425000

 View more versions of this product

Catalog No. AC147425000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95%
Formula Weight 161.49
Quantity 500mL
Color Clear colorless to slightly brown
EINECS Number 200-917-8
TSCA TSCA
Density 1.2700g/mL
RTECS Number VV6125000
Chemical Name or Material Trichlorovinylsilane
Infrared Spectrum Authentic
Synonym VTC
Health Hazard 1 Danger
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 96%
Assay Percent Range 96%
Molecular Formula C2H3Cl3Si
Linear Formula H2C=CHSiCl3
MDL Number MFCD00000482
CAS 75-94-5

Specifications

Chemical Name or Material Trichlorovinylsilane
Name Note 96%
CAS 75-94-5
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C2H3Cl3Si
Linear Formula H2C=CHSiCl3
Formula Weight 161.49
MDL Number MFCD00000482
Synonym VTC
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents