missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α,α,α-Trichlorotoluene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164530050

 View more versions of this product

Catalog No. AC164530050



Specifications

Specifications

α, α, α-Trichlorotoluene
97°C
Glass bottle
1.38
Yellow
5mL
98-07-7
100.0
C7H5Cl3
MFCD00000786
15, 1112
Solubility in water: hydrolysis. Other solubilities: soluble in alcohol,benzene,ether and,most organic solvents
C1=CC=C(C=C1)C(Cl)(Cl)Cl
195.48
CHEBI:82274
Liquid
1.3800g/mL
Authentic
1.5570 to 1.559
219°C to 223°C
-7.5°C
98%
97.5
98%
C6H5CCl3
05, 300
alpha,alpha,alpha-trichlorotoluene
XEMRAKSQROQPBR-UHFFFAOYSA-N
trichloromethylbenzene
7367
195.48
98%

Chemical Identifiers

98-07-7
195.48
XEMRAKSQROQPBR-UHFFFAOYSA-N
7367
trichloromethylbenzene
C7H5Cl3
MFCD00000786
alpha,alpha,alpha-trichlorotoluene
CHEBI:82274
C1=CC=C(C=C1)C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Toxic if inhaled.
May cause cancer.
May cause respiratory irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-634-5

RTECSNumber : XT9275000

TSCA : TSCA

SDS Documents