Please login to your online account to display your discounted pricing

Trichlorosilane, 98%, Alfa Aesar™

$160.08 - $160.08

Chemical Identifiers

Chemical Name or Material Trichlorosilane
CAS 10025-78-2
Molecular Formula HSiCl3
Formula Weight 135.45
MDL Number MFCD00011518
UN Number UN1295
Merck Index 14,9646
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1407836

 
alfa aesar
1407836
Each for $160.08
Description & Specifications

Specifications

Formula Weight 135.45
Quantity 500g
Chemical Name or Material Trichlorosilane
TSCA TSCA
Physical Form Liquid
Refractive Index 1.4020
Assay 98%
Sensitivity Air and Moisture Sensitive
Molecular Formula HSiCl3
Storage Information 2° to 8°C
MDL Number MFCD00011518
CAS 10025-78-2
UN Number UN1295
Merck Index 14,9646
Boiling Point 31° to 32°C
Color Colorless
Melting Point −127°C
Flash Point −13°C (7°F)
Solubility Information Soluble in benzene, CS2, chloroform, CCl4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H250-H314-H302-H332;
Extremely flammable liquid and vapour.
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.342
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: TRICHLOROSILANE
RTECS Number VV5950000
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H250-H314-H302-H332;
Extremely flammable liquid and vapour.
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.