Please login to your online account to display your discounted pricing

Trichloroethylene, Stabilized, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$120.50 - $591.95

Chemical Identifiers

Chemical Name or Material Trichloroethylene
CAS 79-01-6
Name Note Stabilized
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula ClCH:CCl2
Formula Weight 131.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004529

 
avantor performance
945803
Narrow Mouth Amber Glass Bottle 4L Each for $152.38

02004530

 
avantor performance
945805
Aluminum Bottle 4L Each for $591.95

02004531

 
avantor performance
945801
Narrow Mouth Amber Glass Bottle 500mL Each for $120.50
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 131.39
Chemical Name or Material Trichloroethylene
CAS 79-01-6
Molecular Formula ClCH:CCl2
Name Note Stabilized
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.47kg