missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroethylene, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after evaporation 3ppm, H2O 0.02%

Manufacturer:  Alfa Aesar 43487K2

 View more versions of this product

Catalog No. AA43487K2



Specifications

Specifications

Trichloroethylene
>99.5%
MFCD00000838
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
6575
131.39
Sprectrophotometric
410°C (770°F)
58mm Hg at 20°C
1L
79-01-6
C2HCl3
acetylene trichloride
C(=C(Cl)Cl)Cl
131.38
CHEBI:16602
Liquid
1.46g/cm3 at 20°C
Chloroform-like
Colorless

Chemical Identifiers

79-01-6
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
6575
1,1,2-trichloroethene
C2HCl3
MFCD00000838
acetylene trichloride
CHEBI:16602
C(=C(Cl)Cl)Cl
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 201-167-4

Documents