Please login to your online account to display your discounted pricing

Trichloroethylene, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after evaporation 3ppm, H2O 0.02%

$68.65 - $68.65

Chemical Identifiers

Chemical Name or Material Trichloroethylene
CAS 79-01-6
Purity Grade Spectrophotometric grade
Physical Form Liquid
Assay Percent Range >99.5%
Molecular Formula ClCH=CCl2
Formula Weight 131.39
MDL Number MFCD00000838
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43487K2

 
alfa aesar
43487K2
1L Each for $68.65

AA43487K7

 
alfa aesar
43487K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43487M1

 
alfa aesar
43487M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43487M6

 
alfa aesar
43487M6
4 x 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 131.39
Purity Grade Spectrophotometric grade
Color Clear colorless
Quantity 1L
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Trichloroethylene
EINECS Number 201-167-4
Ignition Point 410°C (770°F)
Odor Chloroform-like
Assay Percent Range >99.5%
Molecular Formula ClCH=CCl2
MDL Number MFCD00000838
CAS 79-01-6
Vapor Pressure 58mm Hg at 20°C
Health Hazard 1 H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.
Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
Density 1.46g/cm3 at 20°C
Safety and Handling

H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.