Please login to your online account to display your discounted pricing

Alfa Aesar™ Trichloroethylene, Semiconductor Grade, 99+%

Chemical Identifiers

Chemical Name or Material Trichloroethylene
CAS 79-01-6
Purity Grade Semiconductor Grade
Formula Weight 131.39
MDL Number MFCD00000838
UN Number UN1710
Beilstein 1736782
Merck Index 14,9639
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39744K2

 
alfa aesar™
39744K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39744K7

 
alfa aesar™
39744K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.39
Chemical Name or Material Trichloroethylene
TSCA TSCA
Density 1.46g/mL
Purity Grade Semiconductor Grade
Physical Form Liquid
Assay ≥99%
MDL Number MFCD00000838
CAS 79-01-6
UN Number UN1710
Beilstein 1736782
Merck Index 14,9639
Health Hazard 1 Danger
Odor Chloroform-like
Refractive Index 1.476
Health Hazard 2 GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 87°C
Melting Point −85°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE
RTECS Number KX4550000
Quantity 1L
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.