Please login to your online account to display your discounted pricing

Trichloroethylene ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Trichloroethylene
CAS 79-01-6
Purity Grade ACS Grade
Molecular Formula ClCH:CCl2
Formula Weight 131.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK86004

 
avantor performance
860008*
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.39
Purity Grade ACS Grade
Quantity 4L
Packaging Glass
Molecular Formula ClCH:CCl2
CAS 79-01-6
Chemical Name or Material Trichloroethylene