Learn More
Trichloroethylene, ACS, 99.5% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 019401M1
Description
Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trichloroethylene | |
79-01-6 | |
1.46 g/mL | |
None | |
Chloroform-like | |
1.476 | |
4 x 1 L | |
1736782 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
1,1,2-trichloroethene | |
6575 | |
131.39 | |
ACS |
For ACS analysis | |
-85°C | |
87°C | |
9.5% min. | |
C2HCl3 | |
MFCD00000838 | |
UN1710 | |
14,9639 | |
Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform; Very slightly miscible with water. | |
C(=C(Cl)Cl)Cl | |
131.38 | |
CHEBI:16602 | |
≤99.5% | |
Liquid |
Chemical Identifiers
79-01-6 | |
131.38 | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
6575 | |
1,1,2-trichloroethene |
C2HCl3 | |
MFCD00000838 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
CHEBI:16602 | |
C(=C(Cl)Cl)Cl |
Safety and Handling
GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H336-H341-H350-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE
EINECSNumber : 201-167-4
RTECSNumber : KX4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only