Please login to your online account to display your discounted pricing

Trichloroacetyl isocyanate, NMR grade, ACROS Organics™

$146.05 - $146.05

Chemical Identifiers

Chemical Name or Material Trichloroacetyl isocyanate
Assay ≥97%
Molecular Formula C3Cl3NO2
Linear Formula CCl3CONCO
Formula Weight 188.4
MDL Number MFCD00002033
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC202900020

  SDS
thermo fisher scientific
AC202900020
Glass bottle 2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC202900100

  SDS
thermo fisher scientific
202900100
Glass bottle 10g Each for $146.05

AC202900500

  SDS
thermo fisher scientific
AC202900500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97%
Formula Weight 188.4
Physical Form Liquid
Quantity 2g
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 221-165-7
Density 1.5900g/mL
Refractive Index 1.4799 to 1.4819 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violently with water.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
Chemical Name or Material Trichloroacetyl isocyanate
Linear Formula CCl3CONCO
MDL Number MFCD00002033
Molecular Formula C3Cl3NO2
CAS 3019-71-4

For NMR

Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violently with water.