Learn More
Trichloroacetic anhydride, 95%, Thermo Scientific Chemicals
$154.55 - $577.89
Chemical Identifiers
CAS | 4124-31-6 |
---|---|
Molecular Formula | C4Cl6O3 |
Molecular Weight (g/mol) | 308.741 |
MDL Number | MFCD00000793 |
InChI Key | MEFKFJOEVLUFAY-UHFFFAOYSA-N |
Synonym | trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate |
PubChem CID | 20079 |
IUPAC Name | (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate |
SMILES | C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0084918
|
Thermo Scientific Chemicals
L0084918 |
50 g |
Each for $154.55
|
|
|||||
AAL0084930
|
Thermo Scientific Chemicals
L0084930 |
250 g |
Each for $577.89
|
|
|||||
Description
Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.
Solubility
Miscible with ether and acetic acid.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases with strong oxidizing agents.
Chemical Identifiers
4124-31-6 | |
308.741 | |
MEFKFJOEVLUFAY-UHFFFAOYSA-N | |
20079 | |
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl |
C4Cl6O3 | |
MFCD00000793 | |
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate | |
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate |
Specifications
4124-31-6 | |
98°C to 100°C (11 mmHg) | |
1.484 | |
50 g | |
980355 | |
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate | |
MEFKFJOEVLUFAY-UHFFFAOYSA-N | |
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate | |
20079 | |
95% |
1.69 | |
C4Cl6O3 | |
MFCD00000793 | |
UN3265 | |
Moisture sensitive | |
Miscible with ether and acetic acid. | |
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl | |
308.741 | |
308.76 | |
Trichloroacetic anhydride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 223-925-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only