missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trichloroacetic anhydride, 95%

Manufacturer:  Alfa Aesar™ L0084930

 View more versions of this product

Catalog No. AAL0084930



Description

Description

Applications
Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.

Solubility
Miscible with ether and acetic acid.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases with strong oxidizing agents.
Specifications

Specifications

1.69
Moisture sensitive
250g
C4Cl6O3
UN3265
2,2,2-trichloroacetic anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acetic acid, trichloro-, anhydride, acmc-1aliy, bis trichloroacetic anhydride, perchloroacetic anhydride, trichloroacetic acid anhydride, trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
MEFKFJOEVLUFAY-UHFFFAOYSA-N
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
20079
95%
1.484
98°C to 100°C (11mmHg)
4124-31-6
MFCD00000793
980355
Miscible with ether and acetic acid.
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
308.741
308.76

Chemical Identifiers

4124-31-6
308.741
MEFKFJOEVLUFAY-UHFFFAOYSA-N
20079
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
C4Cl6O3
MFCD00000793
2,2,2-trichloroacetic anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acetic acid, trichloro-, anhydride, acmc-1aliy, bis trichloroacetic anhydride, perchloroacetic anhydride, trichloroacetic acid anhydride, trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 223-925-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents