Learn More
Trichloroacetic acid, sodium salt, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 184155000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trichloroacetic acid, sodium salt | |
97% | |
CCl3CO2Na | |
01,1107; 02,388; 06,553; 07,341 | |
sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca | |
C(=O)(C(Cl)(Cl)Cl)[O-].[Na+] | |
185.36 | |
CHEBI:81976 | |
97% | |
Authentic |
650-51-1 | |
C2Cl3NaO2 | |
MFCD00064198 | |
15, 9792 | |
SAQSTQBVENFSKT-UHFFFAOYSA-M | |
sodium;2,2,2-trichloroacetate | |
23681045 | |
185.37 | |
500g | |
Plastic bottle |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 211-479-2
RTECSNumber : AJ9100000
TSCA : TSCA
RUO â Research Use Only