Learn More
Trichloroacetic Acid (Flakes or Crystals), Fisher BioReagents
$113.76 - $254.11

Chemical Identifiers
CAS | 76-03-9 |
---|---|
Molecular Formula | C2HCl3O2 |
Molecular Weight (g/mol) | 163.378 |
InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
Synonym | acetic acid, trichloro, aceto-caustin, acide trichloracetique, acido tricloroacetico, amchem grass killer, konesta, trichloracetic acid, trichloressigsaeure, trichloroacetic acid, trichloroethanoic acid |
PubChem CID | 6421 |
ChEBI | CHEBI:30956 |
IUPAC Name | 2,2,2-trichloroacetic acid |
SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP555-250
|
Fisher BioReagents
FLBP555250 |
250g | Glass Bottle |
Each for $113.76
Case of 6 Each for $682.56 |
|
BP555-500
|
Fisher BioReagents
FLBP555500 |
500g | Glass Bottle |
Each for $157.24
Case of 6 Each for $943.44 |
|
BP555-1
|
Fisher BioReagents
FLBP5551 |
1kg | Glass Bottle |
Each for $254.11
Case of 4 Each for $1,016.44 |
|
Description
TCA is used to precipitate proteins and nucleic acids from solution.Chemical Identifiers
76-03-9 | |
163.378 | |
acetic acid, trichloro, aceto-caustin, acide trichloracetique, acido tricloroacetico, amchem grass killer, konesta, trichloracetic acid, trichloressigsaeure, trichloroacetic acid, trichloroethanoic acid | |
CHEBI:30956 | |
C(=O)(C(Cl)(Cl)Cl)O |
C2HCl3O2 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
6421 | |
2,2,2-trichloroacetic acid |
Specifications
76-03-9 | |
≥99.0 % | |
CCl3COOH | |
1839 | |
acetic acid, trichloro, aceto-caustin, acide trichloracetique, acido tricloroacetico, amchem grass killer, konesta, trichloracetic acid, trichloressigsaeure, trichloroacetic acid, trichloroethanoic acid | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
2,2,2-trichloroacetic acid | |
6421 | |
163.39 | |
≥99.0% | |
Pass Test | |
0.001% max. | |
Glass Bottle | |
Pass Test | |
1.2mbar at 50°C, 0.08mbar at 25°C | |
1.2 | |
196°C | |
Trichloroacetic Acid |
Vapor Pressure: 1.2mbar at 50°C | |
C2HCl3O2 | |
0.002% max. | |
15, 9792 | |
1.620g/cm³ | |
C(=O)(C(Cl)(Cl)Cl)O | |
163.378 | |
CHEBI:30956 | |
Solid | |
0.002% max. | |
0.030% max. | |
0.002% max. | |
5ppm max. | |
0.02% max. | |
52°C | |
White | |
250g |
Safety and Handling

DANGER!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:3
Flammability:1
Instability:1
DOTInformation : DOT Class 8, : Corrosive
Recommended Storage : RT