missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroacetic Acid, Certified, 80.0% (w/v) ±2.0%, LabChem™

$211.11 - $211.11

Chemical Identifiers

CAS 76-03-9,7732-18-5
CAS Max % 80,20
Linear Formula C2HCl3O2
Formula Weight 163.39
UN Number UN2564
Synonym TCA
Chemical Name or Material Trichloroacetic Acid
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC262651 Lab Chem
LC262651
500mL Each for $211.11
Specifications Description & Specifications

Chemical Identifiers

76-03-9,7732-18-5
C2HCl3O2
UN2564
Trichloroacetic Acid
80,20
163.39
TCA
Certified Grade

Specifications

163.39
Chlorine; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
UN2564
20,80
Traceable to NIST
7732-18-5,76-03-9
Amber Glass
Trichloroacetic Acid
Certified Grade
Clear, colorless to slight tan
TCA
500mL
Soluble in water
Liquid
C2HCl3O2
Passes Test
80.0% ± 2.0% (w/v)

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer (oral).

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Danger

Recommended Storage : Room Temperature