Please login to your online account to display your discounted pricing

Trichloroacetic Acid, Certified, 5.0% (w/v) ±0.4%, LabChem™

$54.89 - $54.89

Chemical Identifiers

Chemical Name or Material Trichloroacetic Acid
CAS 7732-18-5
CAS Max % 95
CAS 76-03-9
CAS Max % 5
Purity Grade Certified Grade
Linear Formula C2HCl3O2
Formula Weight 163.39
UN Number UN2564
Synonym TCA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC262201

 
lab chem
LC262201
500mL Each for $54.89
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 5.0% ± 0.4% (w/v)
Formula Weight 163.39
CAS Max % 5
Solubility Soluble in water
Synonym TCA
Packaging Amber Glass
Chemical Name or Material Trichloroacetic Acid
UN Number UN2564
Quantity 500mL
Health Hazard 1 Danger
CAS 7732-18-5
Traceability to NIST Traceable to NIST
CAS Max % 95
CAS 76-03-9
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Decomposition Information Chlorine; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Identification Passes Test
Physical Form Liquid
Color Clear, colorless
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer (oral).
Linear Formula C2HCl3O2
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer (oral).

Room Temperature