Please login to your online account to display your discounted pricing

Trichloroacetic Acid, Certified, 3.0% (w/v) ±0.3%, LabChem™

$70.60 - $70.60

Chemical Identifiers

Chemical Name or Material Trichloroacetic Acid
CAS 7732-18-5
CAS Max % 97
CAS 76-03-9
CAS Max % 3
Purity Grade Certified Grade
Linear Formula C2HCl3O2
Formula Weight 163.39
UN Number UN2564
Synonym TCA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC262001

 
lab chem
LC262001
500mL Each for $70.60
Description & Specifications

Specifications

Formula Weight 163.39
Purity Grade Certified Grade
Assay 3.0% ± 0.3% (w/v)
Recommended Storage Room Temperature
Solubility Soluble in water
Synonym TCA
Color Clear, colorless
Packaging Amber Glass
Health Hazard 1 Danger
Identification Passes Test
UN Number UN2564
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Suspected of causing cancer (oral).
CAS 76-03-9
Physical Form Liquid
CAS 7732-18-5
CAS Max % 3
CAS Max % 97
Quantity 500mL
Linear Formula C2HCl3O2
Chemical Name or Material Trichloroacetic Acid
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Traceability to NIST Traceable to NIST
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Suspected of causing cancer (oral).

Room Temperature