Please login to your online account to display your discounted pricing

Trichloroacetic Acid ACS, EMD Millipore


Quantity 500g
Packaging Glass Bottle

Manufacturer: emd millipore corporation  TX1045-1

 View more versions of this product

Catalog No. MTX10451

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Glass Bottle
Quantity 500g
Chemical Name or Material Trichloroacetic Acid
CAS 76-03-9
Purity Grade ACS
Linear Formula CCl3COOH
Formula Weight 163.39g/mol

Specifications

Chemical Name or Material Trichloroacetic Acid
CAS 76-03-9
Purity Grade ACS
Linear Formula CCl3COOH
Formula Weight 163.39g/mol