Please login to your online account to display your discounted pricing

Trichloroacetic Acid ACS, EMD Millipore

$289.09 - $289.09

Chemical Identifiers

Chemical Name or Material Trichloroacetic Acid
CAS 76-03-9
Purity Grade ACS
Linear Formula CCl3COOH
Formula Weight 163.39g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MTX10451

 
emd millipore corporation
TX1045-1
Glass Bottle 500g Each for $289.09
Description & Specifications

Specifications

Packaging Glass Bottle
Quantity 500g
Chemical Name or Material Trichloroacetic Acid
CAS 76-03-9
Purity Grade ACS
Linear Formula CCl3COOH
Formula Weight 163.39g/mol