missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroacetic acid, ACS, 99%, Alfa Aesar™

Protein precipitation reagent

Manufacturer:  Alfa Aesar 2215618

 View more versions of this product

Catalog No. AA2215618



Specifications

Specifications

Trichloroacetic acid
76-03-9
C2HCl3O2
UN1839
14,9627
1.629
C(=O)(C(Cl)(Cl)Cl)O
163.378
CHEBI:30956
Crystalline
ACS
For ACS analysis
99%
MFCD00004177
970119
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
YNJBWRMUSHSURL-UHFFFAOYSA-N
2,2,2-trichloroacetic acid
6421
163.39
99%
50g

Chemical Identifiers

76-03-9
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
6421
2,2,2-trichloroacetic acid
C2HCl3O2
MFCD00004177
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
CHEBI:30956
C(=O)(C(Cl)(Cl)Cl)O
Safety and Handling

Safety and Handling

GHS H Statement
H314

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P363-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETIC ACID

RTECSNumber : AJ7875000

TSCA : TSCA

SDS Documents