Please login to your online account to display your discounted pricing

Trichloroacetic Acid, 33% (w/v) Aqueous Solution, Ricca Chemical

$89.64 - $89.64

Chemical Identifiers

Chemical Name or Material Trichloroacetic Acid
CAS 7732-18-5
CAS Min % 74.0
CAS Max % 77.0
CAS 76-03-9
CAS Min % 24.29
CAS Max % 25.28
CAS 7440-44-0
CAS Min % 0.15
CAS Max % 0.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

868834

 
ricca chemical company
RI8688.34
Amber Glass 120mL Each for $89.64

86883-1

 
ricca chemical company
RI8688.31
Amber Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8688316

 
ricca chemical company
RI8688.316
Amber Glass Bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8688332

 
ricca chemical company
RI8688.332
Amber Glass Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 120mL
Chemical Name or Material Trichloroacetic Acid
Shelf Life 12 months
CAS Max % 0.15
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 33% (w/v) Aqueous Solution
CAS Min % 74.0
CAS Max % 77.0
CAS 76-03-9
CAS Min % 24.29
Packaging Amber Glass
CAS Max % 25.28
CAS 7440-44-0
CAS Min % 0.15
Safety and Handling

12 months