Please login to your online account to display your discounted pricing

Trichloroacetic Acid, 10% w/v (AOAC), Fisher Chemical

$127.55 - $127.55

Chemical Identifiers

Chemical Name or Material Trichloroacetic Acid
Name Note 10% (w/v)
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 76-03-9
CAS Min % 8.9
CAS Max % 9.26
CAS 7440-44-0
CAS Min % 0.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA9410-1

 
ricca chemical company
RISA94101
Amber Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SA9410-500

  SDS
ricca chemical company
RISA9410500
Amber Glass 500mL Each for $127.55
Description & Specifications

Specifications

Quantity 1L
DOT Information DOT Class 8, : Corrosive
Methods For AOAC Method 953.08
NIST Traceable to NIST.
Chemical Name or Material Trichloroacetic Acid
CAS Max % 0.19
Name Note 10% (w/v)
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS Min % 89.0
CAS Max % 93.0
CAS Min % 8.9
Packaging Amber Glass
CAS Min % 0.18
CAS 76-03-9
CAS Max % 9.26
CAS 7440-44-0
Safety and Handling