missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trichloro(6-phenylhexyl)silane 98.0+%, TCI America™
$318.52 - $1113.19
Chemical Identifiers
CAS | 18035-33-1 |
---|---|
Molecular Formula | C12H17Cl3Si |
Molecular Weight (g/mol) | 295.70 |
MDL Number | MFCD09953885 |
InChI Key | ODIAUNNUTSSHMI-UHFFFAOYSA-N |
Synonym | (6-Phenylhexyl)trichlorosilane |
PubChem CID | 53427322 |
IUPAC Name | trichloro(6-phenylhexyl)silane |
SMILES | Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1 |
Chemical Identifiers
18035-33-1 | |
295.70 | |
ODIAUNNUTSSHMI-UHFFFAOYSA-N | |
53427322 | |
Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1 |
C12H17Cl3Si | |
MFCD09953885 | |
(6-Phenylhexyl)trichlorosilane | |
trichloro(6-phenylhexyl)silane |
Specifications
18035-33-1 | |
92°C | |
MFCD09953885 | |
2987 | |
ODIAUNNUTSSHMI-UHFFFAOYSA-N | |
trichloro(6-phenylhexyl)silane | |
53427322 | |
≥98.0% (GC) | |
Trichloro(6-phenylhexyl)silane |
Colorless | |
C12H17Cl3Si | |
5 g | |
(6-Phenylhexyl)trichlorosilane | |
Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1 | |
295.70 | |
295.70 | |
Liquid |
Safety and Handling
TSCA : No