missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloro(3-phenylpropyl)silane 97.0+%, TCI America™
Supplier: TCI America T285325G
Specifications
| Trichloro(3-phenylpropyl)silane | |
| Colorless | |
| C9H11Cl3Si | |
| 25 g | |
| (3-Phenylpropyl)trichlorosilane | |
| C1=CC=C(C=C1)CCC[Si](Cl)(Cl)Cl | |
| 253.622 | |
| 253.62 | |
| Liquid |
| 13617-40-8 | |
| 126°C | |
| MFCD09953886 | |
| 2987 | |
| QJOHXSVBLYVERP-UHFFFAOYSA-N | |
| trichloro(3-phenylpropyl)silane | |
| 12441988 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 13617-40-8 | |
| 253.622 | |
| QJOHXSVBLYVERP-UHFFFAOYSA-N | |
| 12441988 | |
| C1=CC=C(C=C1)CCC[Si](Cl)(Cl)Cl |
| C9H11Cl3Si | |
| MFCD09953886 | |
| (3-Phenylpropyl)trichlorosilane | |
| trichloro(3-phenylpropyl)silane |
Safety and Handling
TSCA : No