missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloro(1H,1H,2H,2H-tridecafluoro-n-octyl)silane, TCI America™

Manufacturer:  TCI America T257725G

 View more versions of this product

Catalog No. T257725G



Specifications

Specifications

Trichloro(1H,1H,2H,2H-tridecafluoro-n-octyl)silane
Colorless
78560-45-9
MFCD00042363
trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane
C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
481.534
481.53
≥97.0% (GC)
85°C
25g
C8H4Cl3F13Si
2986
PISDRBMXQBSCIP-UHFFFAOYSA-N
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
123578
Clear Liquid at 20°C

Chemical Identifiers

78560-45-9
481.534
PISDRBMXQBSCIP-UHFFFAOYSA-N
123578
C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C8H4Cl3F13Si
MFCD00042363
trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2046

TSCA : No

Documents