Learn More
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139360050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tricarballylic acid | |
99-14-9 | |
White to Brown | |
99% | |
C6H8O6 | |
MFCD00002723 | |
02, 815 | |
tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid | |
OC(=O)CC(CC(O)=O)C(O)=O | |
176.12 | |
CHEBI:45969 | |
99% |
99% | |
154°C to 159°C | |
Authentic | |
Glass bottle | |
HO2CCH2CH(CO2H)CH2CO2H | |
5 g | |
15, 9787 | |
KQTIIICEAUMSDG-UHFFFAOYSA-N | |
propane-1,2,3-tricarboxylic acid | |
14925 | |
176.13 | |
Fine Crystalline Powder |
Chemical Identifiers
99-14-9 | |
176.12 | |
KQTIIICEAUMSDG-UHFFFAOYSA-N | |
14925 | |
propane-1,2,3-tricarboxylic acid |
C6H8O6 | |
MFCD00002723 | |
tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid | |
CHEBI:45969 | |
OC(=O)CC(CC(O)=O)C(O)=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 202-733-3
TSCA : TSCA
RUO – Research Use Only