missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tributylphenyltin, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC370230500

 View more versions of this product

Catalog No. AC370230500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Tributylphenyltin
>110°C
Glass bottle
1.125
Colorless
960-16-7
C6H5Sn[(CH2)3CH3]3
phenyl tnbutyl tin, phenyl tributyl tin, phenyltri-n-butyltin, phenyltributlytin, phenyltributylstannane, phenyltributyltin, stannane, tributylphenyl, tributyl phenyl stannane, tributylphenyl tin, tributylphenyltin
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
367.16
367.16
≥96.0%
1.1250g/mL
Authentic
1.5150 to 1.5170
125.0°C to 128.0°C (0.1 mmHg)
50g
C18H32Sn
MFCD00134394
SYUVAXDZVWPKSI-UHFFFAOYSA-N
tributyl(phenyl)stannane
607632
Liquid

Chemical Identifiers

960-16-7
367.16
SYUVAXDZVWPKSI-UHFFFAOYSA-N
607632
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
C18H32Sn
MFCD00134394
phenyl tnbutyl tin, phenyl tributyl tin, phenyltri-n-butyltin, phenyltributlytin, phenyltributylstannane, phenyltributyltin, stannane, tributylphenyl, tributyl phenyl stannane, tributylphenyl tin, tributylphenyltin
tributyl(phenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for se

GHS Signal Word: Danger

SDS Documents