missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tributylphenyltin, ACROS Organics™

Manufacturer:  Acros Organics AC370230010

 View more versions of this product

Catalog No. AC370230010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tributylphenyltin
>110°C
Glass bottle
1.125
Colorless
960-16-7
C6H5Sn[(CH2)3CH3]3
phenyl tnbutyl tin
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
367.164
367.16
≥96.0%
1.1250g/mL
Authentic
1.5150 to 1.5170
125.0°C to 128.0°C (0.1 mmHg)
1g
C18H32Sn
MFCD00134394
SYUVAXDZVWPKSI-UHFFFAOYSA-N
tributyl(phenyl)stannane
607632
Liquid

Chemical Identifiers

960-16-7
367.164
SYUVAXDZVWPKSI-UHFFFAOYSA-N
607632
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
C18H32Sn
MFCD00134394
phenyl tnbutyl tin
tributyl(phenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents