Please login to your online account to display your discounted pricing

Tributylphenyltin, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370230100

 View more versions of this product

Catalog No. AC370230100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 367.16
Quantity 10g
Color Clear colorless
Density 1.1250g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Chemical Name or Material Tributylphenyltin
MDL Number MFCD00134394
Health Hazard 2 GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.
Linear Formula C6H5Sn[(CH2)3CH3]3
Health Hazard 1 Danger
CAS 960-16-7
Molecular Formula C18H32Sn
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tributylphenyltin
Assay ≥96.0%
Molecular Formula C18H32Sn
Linear Formula C6H5Sn[(CH2)3CH3]3
Formula Weight 367.16
MDL Number MFCD00134394
Accessories

Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

SDS & Safety Documents