Please login to your online account to display your discounted pricing

Tributylborane, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC177101000

 View more versions of this product

Catalog No. AC177101000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 182.16
Quantity 100mL
Color Clear colorless to light yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
EINECS Number 204-554-6
TSCA TSCA
Concentration 0.94 to 1.06M
RTECS Number ED1850000
Density 0.8470g/mL
Fieser 10,410
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides.
CAS 109-99-9
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se
Chemical Name or Material Tributylborane
Name Note 1M Solution in Tetrahydrofuran
Molecular Formula C12H27B
Linear Formula [CH3(CH2)3]3B
MDL Number MFCD00009423
CAS 122-56-5

Specifications

Chemical Name or Material Tributylborane
Name Note 1M Solution in Tetrahydrofuran
CAS 122-56-5
CAS 109-99-9
Molecular Formula C12H27B
Linear Formula [CH3(CH2)3]3B
Formula Weight 182.16
MDL Number MFCD00009423
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se

GHS P Statement Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides.

SDS & Safety Documents