Learn More
Tributylborane, 1M solution in THF, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 177108000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
Tributylborane | |
122-56-5,109-99-9 | |
[CH3(CH2)3]3B | |
0.94 to 1.06M | |
0.8470g/mL | |
CMHHITPYCHHOGT-UHFFFAOYSA-N | |
tributylborane | |
31216 | |
AcroSeal™ Glass Bottle | |
800mL |
1M Solution in Tetrahydrofuran | |
C12H27B | |
MFCD00009423 | |
10,410 | |
tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane | |
CCCCB(CCCC)CCCC | |
182.16 | |
182.16 | |
0.847 | |
−20°C |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides. Catches fire spontaneously if exposed to air. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
EINECSNumber : 204-554-6
RTECSNumber : ED1850000
TSCA : TSCA
RUO â Research Use Only