Learn More
Tributylamine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139320025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tributylamine | |
0.7780g/mL | |
75°C | |
98.5% min. (GC) | |
C12H27N | |
[CH3NCH2)3]3N | |
2.5L | |
01,1189; 06,601 | |
15,978 | |
Solubility in water: 0.08g/L water (20°C). Other solubilities: soluble in most organic solvents,very soluble in alcohol and ether | |
CCCCN(CCCC)CCCC | |
185.35 | |
2.4 mPa.s (20°C) | |
185.35 |
102-82-9 | |
215.0°C | |
Authentic | |
Glass bottle | |
1.4273 to 1.4293 | |
MFCD00009431 | |
04,157 | |
0.778 | |
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 | |
IMFACGCPASFAPR-UHFFFAOYSA-N | |
N,N-dibutylbutan-1-amine | |
7622 | |
CHEBI:38905 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position c
GHS Signal Word: Danger
EINECSNumber : 203-058-7
RUO â Research Use Only