Please login to your online account to display your discounted pricing

Tributyl(vinyl)tin, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350000010

 View more versions of this product

Catalog No. AC350000010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 317.09
Quantity 1g
Color Clear colorless to pale yellow
Density 1.0800g/mL
Infrared Spectrum Authentic
Refractive Index 1.4770 to 1.4790 (20°C, 589nm)
Molecular Formula C14H30Sn
Chemical Name or Material Tributyl(vinyl)tin
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00009421
CAS 7486-35-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tributyl(vinyl)tin
CAS 7486-35-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H30Sn
Formula Weight 317.09
MDL Number MFCD00009421
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

SDS & Safety Documents