Please login to your online account to display your discounted pricing

Tributyl phosphite, 94%, ACROS Organics™

$152.87 - $152.87

Chemical Identifiers

Chemical Name or Material Tributyl phosphite
Name Note 94%
CAS 102-85-2
CAS 71-36-3
Assay 92% min.
Assay Percent Range 94%
Molecular Formula C12H27O3P
Linear Formula {CH3(CH2)3O}3P
Formula Weight 250.32g/mol
MDL Number MFCD00009437
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC169920010

  SDS
thermo fisher scientific
169920010
Glass bottle 1kg Each for $152.87
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Density 0.915g/mL
Color Clear colorless
Quantity 1kg
Chemical Name or Material Tributyl phosphite
Name Note 94%
CAS 102-85-2
CAS 71-36-3
Physical Form Clear colorless liquid
Assay 92% min.
Assay Percent Range 94%
Molecular Formula C12H27O3P
Linear Formula {CH3(CH2)3O}3P
Formula Weight 250.32g/mol
MDL Number MFCD00009437
Beilstein 01, I, 187
Fieser 15,280
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number TH0875000
EINECS Number 203-061-3
TSCA TSCA
Safety and Handling

MOISTURE SENSITIVE

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.