missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tributyl borate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 177191000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tributyl borate | |
-70.0°C | |
0.8530g/mL | |
93°C | |
98% | |
C12H27BO3 | |
1.4080 to 1.4100 | |
0.853 | |
LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
tributyl borate | |
12712 | |
98% |
688-74-4 | |
Colorless | |
230.0°C to 235.0°C | |
Authentic | |
Glass bottle | |
(CH3CH2CH2CH2O)3B | |
100 mL | |
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
B(OCCCC)(OCCCC)OCCCC | |
230.155 | |
230.15 | |
Liquid |
Chemical Identifiers
688-74-4 | |
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
tributyl borate |
LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
12712 | |
B(OCCCC)(OCCCC)OCCCC |
RUO – Research Use Only