Please login to your online account to display your discounted pricing

Tributyl(3-methyl-2-butenyl)tin, 90%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370220010

 View more versions of this product

Catalog No. AC370220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88%
Formula Weight 359.18
Physical Form Liquid
Quantity 1g
Color Clear colorless
Density 1.0690g/mL
Refractive Index 1.4860 to 1.4900 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Tributyl(3-methyl-2-butenyl)tin
Molecular Formula C17H36Sn
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula [CH3(CH2)3]3SnCH2CH=C(CH3)2
CAS 53911-92-5
Assay Percent Range 90%
Name Note 90%
MDL Number MFCD01863649

Specifications

Chemical Name or Material Tributyl(3-methyl-2-butenyl)tin
Name Note 90%
CAS 53911-92-5
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C17H36Sn
Linear Formula [CH3(CH2)3]3SnCH2CH=C(CH3)2
Formula Weight 359.18
MDL Number MFCD01863649
Accessories

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

SDS & Safety Documents