missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tributyl(3-methyl-2-butenyl)tin, 90%, ACROS Organics™

Chemical Identifiers

CAS 53911-92-5
Assay Percent Range 90%
Molecular Formula C17H36Sn
Linear Formula [CH3(CH2)3]3SnCH2CH=C(CH3)2
Formula Weight 359.18
MDL Number MFCD01863649
Chemical Name or Material Tributyl(3-methyl-2-butenyl)tin, 90%
Assay ≥88%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC370220050 SDS Thermo Fisher Scientific
AC370220050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC370220010 SDS Thermo Fisher Scientific
AC370220010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

53911-92-5
C17H36Sn
359.18
Tributyl(3-methyl-2-butenyl)tin, 90%
90%
[CH3(CH2)3]3SnCH2CH=C(CH3)2
MFCD01863649
≥88%

Specifications

53911-92-5
Glass bottle
Liquid
C17H36Sn
359.18
1.0690g/mL
5g
≥88%
Authentic
1.4860 to 1.4900 (20°C, 589nm)
90%
[CH3(CH2)3]3SnCH2CH=C(CH3)2
MFCD01863649
Clear colorless
Tributyl(3-methyl-2-butenyl)tin, 90%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Toxic if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger