Please login to your online account to display your discounted pricing

α,α,α-Tribromoquinaldine, 98+%, ACROS Organics™


Quantity 10g

Manufacturer: thermo fisher scientific  174150100

 View more versions of this product

Catalog No. AC174150100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 379.88
Physical Form Crystalline Powder
Quantity 10g
Color Cream or orange-brown
TSCA TSCA
RTECS Number VC3250750
EINECS Number 210-347-1
Melting Point 127° to 131°C
Beilstein 20, II, 243
CAS Max % 100.0
CAS Min % 98.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H6Br3N
MDL Number MFCD00006755
Assay Percent Range 98+%
Name Note 98+%
CAS 613-53-6
Chemical Name or Material α, α, α-Tribromoquinaldine

Specifications

Chemical Name or Material α, α, α-Tribromoquinaldine
Name Note 98+%
CAS 613-53-6
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C10H6Br3N
Formula Weight 379.88
MDL Number MFCD00006755
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents