Please login to your online account to display your discounted pricing

α,α,α-Tribromoquinaldine, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α, α, α-Tribromoquinaldine
Name Note 98+%
CAS 613-53-6
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C10H6Br3N
Formula Weight 379.88
MDL Number MFCD00006755
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC174150100

  SDS
thermo fisher scientific
174150100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 379.88
Quantity 10g
Color Cream or orange-brown
TSCA TSCA
RTECS Number VC3250750
EINECS Number 210-347-1
Melting Point 127° to 131°C
Beilstein 20, II, 243
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material α, α, α-Tribromoquinaldine
Infrared Spectrum Authentic
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C10H6Br3N
MDL Number MFCD00006755
CAS 613-53-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.