Please login to your online account to display your discounted pricing

Tribromoacetic Acid 99%, ACROS Organics™

$87.17 - $87.17

Chemical Identifiers

Chemical Name or Material Tribromoacetic acid
Name Note 99%
CAS 75-96-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2HBr3O2
Linear Formula CBr3CO2H
Formula Weight 296.73
MDL Number MFCD00004167
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139300050

  SDS
thermo fisher scientific
AC139300050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC139300250

  SDS
thermo fisher scientific
139300250
Glass bottle 25g Each for $87.17
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 296.73
Physical Form Crystals
Quantity 5g
Color White to light yellow
EINECS Number 200-919-9
TSCA TSCA
Melting Point 130° to 133°C
Beilstein 02, 220
Merck Index 15, 9770
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Linear Formula CBr3CO2H
Name Note 99%
Molecular Formula C2HBr3O2
CAS 75-96-7
Chemical Name or Material Tribromoacetic acid
MDL Number MFCD00004167
Assay Percent Range 99%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.