Please login to your online account to display your discounted pricing

Alfa Aesar™ Tribromoacetic acid, 97%

$41.24 - $41.24

Chemical Identifiers

Chemical Name or Material Tribromoacetic acid
CAS 75-96-7
Molecular Formula CBr3CO2H
Formula Weight 296.74
MDL Number MFCD00004167
UN Number UN3261
Beilstein 1755150
Merck Index 14,9609
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2436706

 
alfa aesar™
B2436706
5g Each for $41.24

AAB2436714

 
alfa aesar™
B2436714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.74
Chemical Name or Material Tribromoacetic acid
TSCA TSCA
Assay 97%
Molecular Formula CBr3CO2H
MDL Number MFCD00004167
CAS 75-96-7
UN Number UN3261
Beilstein 1755150
Merck Index 14,9609
Melting Point 130° to 133°C
Boiling Point 244° to 246°C
Health Hazard 1 Danger
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.