missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tribenzyltin chloride

Manufacturer:  Alfa Aesar™ 7120114

 View more versions of this product

Catalog No. AA7120114



Specifications

Specifications

Tribenzyltin chloride
3151-41-5
Moisture Sensitive
MFCD00014431
PUFMEHZUXSPPAM-UHFFFAOYSA-M
tribenzyl(chloro)stannane
18470
Powder
143°C
25g
C21H21ClSn
UN3146
C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl
427.559
427.54

Chemical Identifiers

3151-41-5
427.559
PUFMEHZUXSPPAM-UHFFFAOYSA-M
tribenzyl(chloro)stannane
C21H21ClSn
MFCD00014431
18470
C1=CC=C(C=C1)C[Sn](CC2=CC=CC=C2)(CC3=CC=CC=C3)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319-H335
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

SDS Documents