missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tribenzylphosphine

Manufacturer:  Alfa Aesar™ 1017303

 View more versions of this product

Catalog No. AA1017303



Specifications

Specifications

Tribenzylphosphine
183°C to 185°C (0.2mmHg)
93°C to 99°C
7650-89-7
(C6H5CH2)3P
acmc-20ajfj, phosphine, tribenzyl, phosphine, tris phenylmethyl, tribenzyl phosphine, tribenzylphosphine, tribenzylphosphine 1g, tris phenylmethyl phosphane, tris phenylmethyl phosphine
IFXORIIYQORRMJ-UHFFFAOYSA-N
tribenzylphosphane
82101
Crystalline
Air and moisture sensitive
White
1g
C21H21P
MFCD00014439
Insoluble in water.
C1=CC=C(C=C1)CP(CC2=CC=CC=C2)CC3=CC=CC=C3
304.373
304.37

Chemical Identifiers

7650-89-7
304.373
IFXORIIYQORRMJ-UHFFFAOYSA-N
82101
C1=CC=C(C=C1)CP(CC2=CC=CC=C2)CC3=CC=CC=C3
C21H21P
MFCD00014439
acmc-20ajfj, phosphine, tribenzyl, phosphine, tris phenylmethyl, tribenzyl phosphine, tribenzylphosphine, tribenzylphosphine 1g, tris phenylmethyl phosphane, tris phenylmethyl phosphine
tribenzylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-608-6

RTECSNumber : SZ3865000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents