missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tribenzylphosphine

Manufacturer:  Alfa Aesar™ 1017306

 View more versions of this product

Catalog No. AA1017306



Specifications

Specifications

Tribenzylphosphine
183°C to 185°C (0.2mmHg)
93°C to 99°C
7650-89-7
(C6H5CH2)3P
acmc-20ajfj
C1=CC=C(C=C1)CP(CC2=CC=CC=C2)CC3=CC=CC=C3
304.373
304.37
Air & Moisture Sensitive
White
5g
C21H21P
MFCD00014439
IFXORIIYQORRMJ-UHFFFAOYSA-N
tribenzylphosphane
82101
Crystalline

Chemical Identifiers

7650-89-7
304.373
IFXORIIYQORRMJ-UHFFFAOYSA-N
82101
C1=CC=C(C=C1)CP(CC2=CC=CC=C2)CC3=CC=CC=C3
C21H21P
MFCD00014439
acmc-20ajfj
tribenzylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SZ3865000

TSCA : TSCA

Documents