missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tribenzylamine, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 156470050

 View more versions of this product

Catalog No. AC156470050


 

Specifications Description & Specifications

Specifications

≥99.0%
287.39
White to light cream
0.5% max. (K.F.)
620-40-6
MFCD00004773
Tribenzylamine
C21H21N
Powder
5g
90° to 94°C
Authentic
99+%
99+%
(C6H5CH2)3N

Chemical Identifiers

620-40-6
C21H21N
287.39
Tribenzylamine, 99+%
99+%
(C6H5CH2)3N
MFCD00004773
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-638-3

TSCA : TSCA